The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243198.2 | NP_001230127.1 | 13 - 14 | 1249 | 70 |
NM_005338.6 | NP_005329.3 | 13 - 14 | 1249 | 70 | |
XM_005250304.2 | XP_005250361.1 | 13 - 14 | 1250 | 70 | |
XM_005250305.2 | XP_005250362.1 | 13 - 14 | 1255 | 70 | |
XM_011516116.2 | XP_011514418.1 | 13 - 14 | 1253 | 70 | |
XM_017012099.1 | XP_016867588.1 | 13 - 14 | 1170 | 70 | |
GenBank mRNA | AF365404.1 | - | 13 - 14 | 1176 | 70 |
AK302382.1 | - | 2 - 3 | 176 | 70 | |
AK304738.1 | - | 13 - 14 | 1218 | 70 | |
BC110545.2 | - | 13 - 14 | 1224 | 70 | |
BC139786.1 | - | 3 - 4 | 303 | 70 | |
KF437291.1 | - | 13 - 14 | 1114 | 70 | |
U79734.1 | - | 12 - 13 | 1072 | 70 | |
Y09420.1 | - | 5 - 6 | 465 | 70 |