The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243198.2 | NP_001230127.1 | 19 - 20 | 2011 | 80 |
NM_005338.6 | NP_005329.3 | 19 - 20 | 2011 | 80 | |
XM_005250304.2 | XP_005250361.1 | 19 - 20 | 2012 | 80 | |
XM_005250305.2 | XP_005250362.1 | 19 - 20 | 2017 | 80 | |
XM_011516116.2 | XP_011514418.1 | 19 - 20 | 2015 | 80 | |
XM_017012099.1 | XP_016867588.1 | 19 - 20 | 1932 | 80 | |
GenBank mRNA | AF365404.1 | - | 19 - 20 | 1938 | 80 |
AK302382.1 | - | 8 - 9 | 938 | 80 | |
AK304738.1 | - | 19 - 20 | 1980 | 80 | |
BC110545.2 | - | 19 - 20 | 1986 | 80 | |
BC139786.1 | - | 9 - 10 | 1065 | 80 | |
KF437291.1 | - | 19 - 20 | 1876 | 80 | |
U79734.1 | - | 18 - 19 | 1834 | 80 |