Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
Chromosome Location
Chr.8: 12945674 - 13029777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005273588.4 XP_005273645.1 1 - 1 118 123
XM_005273591.4 XP_005273648.1 1 - 1 118 123
XM_011544598.2 XP_011542900.1 1 - 1 158 123
XM_017013707.1 XP_016869196.1 1 - 1 118 123
XM_017013714.1 XP_016869203.1 1 - 1 118 123
GenBank mRNA BC035082.1 - 1 - 1 464 123

View Full Product Details