The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006090.3 | NP_006081.1 | 1 - 2 | 101 | 129 |
XM_011540496.1 | XP_011538798.1 | 1 - 2 | 101 | 129 | |
GenBank mRNA | AF068302.1 | - | 1 - 2 | 59 | 129 |
AK023381.1 | - | 1 - 2 | 88 | 129 | |
BX487092.1 | - | 1 - 2 | 101 | 129 |