The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000555.3 | NP_000546.2 | 5 - 6 | 1350 | 91 |
NM_178151.2 | NP_835364.1 | 5 - 6 | 1179 | 91 | |
NM_178153.2 | NP_835366.1 | 5 - 6 | 1052 | 91 | |
XM_011530878.2 | XP_011529180.1 | 5 - 6 | 1024 | 91 | |
GenBank mRNA | AF034634.1 | - | 4 - 5 | 935 | 91 |
AF040255.1 | - | 5 - 6 | 977 | 91 | |
AJ003112.1 | - | 5 - 6 | 1350 | 91 | |
BC027925.1 | - | 5 - 6 | 994 | 91 |