The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204103.1 | NP_001191032.1 | 4 - 5 | 835 | 62 |
NM_005155.6 | NP_005146.4 | 4 - 5 | 845 | 62 | |
NM_138717.2 | NP_619731.2 | 4 - 5 | 624 | 62 | |
GenBank mRNA | AF020543.1 | - | 4 - 5 | 790 | 62 |
AL110128.1 | - | 4 - 5 | 552 | 62 | |
BC001355.2 | - | 4 - 5 | 659 | 62 | |
CR533546.1 | - | 3 - 4 | 449 | 62 | |
Y17958.1 | - | 4 - 5 | 739 | 62 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_037861.1 | 4 - 5 | 845 | 62 | |
GenBank mRNA | AB209572.1 | - | 17 - 18 | 62 | |
AF020544.1 | - | 4 - 5 | 999 | 62 | |
AK290473.1 | - | 4 - 5 | 700 | 62 | |
AK296648.1 | - | 3 - 4 | 620 | 62 | |
AK307427.1 | - | 2 - 3 | 298 | 62 |