The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195386.1 | NP_001182315.1 | 2 - 3 | 178 | 117 |
NM_001195387.1 | NP_001182316.1 | 2 - 3 | 162 | 117 | |
NM_145274.3 | NP_660317.2 | 2 - 3 | 170 | 117 | |
XM_005257089.4 | XP_005257146.1 | 3 - 4 | 367 | 117 | |
XM_017024253.1 | XP_016879742.1 | 3 - 4 | 335 | 117 | |
GenBank mRNA | AK097454.1 | - | 2 - 3 | 111 | 117 |
AK298616.1 | - | 2 - 3 | 113 | 117 | |
BC015365.1 | - | 2 - 3 | 149 | 117 | |
BG530583.1 | - | 2 - 3 | 148 | 117 | |
CD689247.1 | - | 2 - 3 | 195 | 117 | |
DB459363.1 | - | 2 - 3 | 178 | 117 |