Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHOSPHO2
Entrez Gene ID
493911
Gene Name
phosphatase, orphan 2
Gene Aliases
-
Chromosome Location
Chr.2: 169694454 - 169701708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008489.3 NP_001008489.1 3 - 4 370 145
NM_001199285.1 NP_001186214.1 3 - 4 270 145
NM_001199286.1 NP_001186215.1 3 - 4 489 145
GenBank mRNA AK314915.1 - 3 - 4 273 145
BC022324.2 - 2 - 3 201 145
BC106013.1 - 3 - 4 359 145
BI550291.1 - 3 - 4 477 145
DA211528.1 - 3 - 4 273 145

View Full Product Details