Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9
Entrez Gene ID
735
Gene Name
complement component 9
Gene Aliases
ARMD15, C9D
Chromosome Location
Chr.5: 39284275 - 39364553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC020721.1 - 7 - 8 1197 132
K02766.1 - 7 - 8 1102 132
X02176.1 - 7 - 8 1112 132

View Full Product Details