The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030762.2 | NP_110389.1 | 1 - 2 | 351 | 86 |
GenBank mRNA | AB044088.1 | - | 1 - 2 | 194 | 86 |
AK303677.1 | - | 1 - 2 | 190 | 86 | |
AY665467.1 | - | 1 - 2 | 161 | 86 | |
BC025968.1 | - | 1 - 2 | 351 | 86 |