The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002156.4 | NP_002147.2 | 9 - 10 | 1359 | 96 |
NM_199440.1 | NP_955472.1 | 9 - 10 | 1339 | 96 | |
GenBank mRNA | AK297305.1 | - | - | 1286 | 96 |
AK298368.1 | - | 9 - 10 | 1078 | 96 | |
AK298632.1 | - | 9 - 10 | 1282 | 96 | |
AK298844.1 | - | 8 - 9 | 1110 | 96 | |
AK312240.1 | - | 9 - 10 | 1292 | 96 | |
BC002676.2 | - | 9 - 10 | 1269 | 96 | |
BC003030.1 | - | 9 - 10 | 1268 | 96 | |
BC047350.1 | - | 8 - 9 | 1429 | 96 | |
BC067082.1 | - | 9 - 10 | 1267 | 96 | |
BC073746.1 | - | 9 - 10 | 1267 | 96 | |
BX647145.1 | - | 8 - 9 | 2024 | 96 | |
EU719626.1 | - | 8 - 9 | 1222 | 96 | |
KU178139.1 | - | 8 - 9 | 1222 | 96 | |
KU178140.1 | - | 3 - 4 | 473 | 96 | |
M22382.1 | - | 9 - 10 | 1267 | 96 | |
M34664.1 | - | 9 - 10 | 1246 | 96 |