The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201279.1 | NP_958436.1 | 15 - 16 | 3213 | 64 |
GenBank mRNA | AF022859.1 | - | 15 - 16 | 2422 | 64 |