Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP58
Entrez Gene ID
51602
Gene Name
NOP58 ribonucleoprotein
Gene Aliases
HSPC120, NOP5, NOP5/NOP58
Chromosome Location
Chr.2: 202265716 - 202303661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015934.4 NP_057018.1 6 - 7 724 138
GenBank mRNA AF123534.1 - 6 - 7 498 138
AF161469.1 - 4 - 5 380 138
AF263608.1 - 6 - 7 498 138
AK023975.1 - 5 - 6 423 138
AK307396.1 - 6 - 7 704 138
AL117554.1 - 6 - 7 707 138
BC001707.1 - 6 - 7 690 138
BC009306.2 - 6 - 7 654 138
BC018805.1 - 6 - 7 677 138
BC032592.2 - 6 - 7 648 138

View Full Product Details