The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004995.3 | NP_004986.1 | 7 - 8 | 1440 | 67 |
GenBank mRNA | AK291325.1 | - | 7 - 8 | 1382 | 67 |
AK312657.1 | - | 7 - 8 | 1388 | 67 | |
BC064803.1 | - | 7 - 8 | 1374 | 67 | |
D26512.1 | - | 7 - 8 | 1266 | 67 | |
HE978355.1 | - | 7 - 8 | 1389 | 67 | |
U41078.1 | - | 7 - 8 | 1155 | 67 | |
X83535.1 | - | 7 - 8 | 1274 | 67 | |
X90925.1 | - | 7 - 8 | 1267 | 67 | |
Z48481.1 | - | 7 - 8 | 1268 | 67 |