Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK2
Entrez Gene ID
5126
Gene Name
proprotein convertase subtilisin/kexin type 2
Gene Aliases
NEC 2, NEC-2, NEC2, PC2, SPC2
Chromosome Location
Chr.20: 17226107 - 17484578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201528.1 NP_001188457.1 5 - 6 756 88
NM_001201529.2 NP_001188458.1 3 - 4 756 88
NM_002594.4 NP_002585.2 4 - 5 861 88
GenBank mRNA AK294200.1 - 3 - 4 652 88
AK312341.1 - 4 - 5 685 88
BC005815.2 - 4 - 5 762 88
BC040546.1 - 5 - 6 756 88
J05252.1 - 4 - 5 594 88

View Full Product Details