The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201528.1 | NP_001188457.1 | 6 - 7 | 794 | 108 |
NM_001201529.2 | NP_001188458.1 | 4 - 5 | 794 | 108 | |
NM_002594.4 | NP_002585.2 | 5 - 6 | 899 | 108 | |
GenBank mRNA | AK294200.1 | - | 4 - 5 | 690 | 108 |
AK312341.1 | - | 5 - 6 | 723 | 108 | |
BC005815.2 | - | 5 - 6 | 800 | 108 | |
BC040546.1 | - | 6 - 7 | 794 | 108 | |
J05252.1 | - | 5 - 6 | 632 | 108 |