Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMTF1
Entrez Gene ID
9988
Gene Name
cyclin D binding myb like transcription factor 1
Gene Aliases
DMP1, DMTF, MRUL, hDMP1
Chromosome Location
Chr.7: 87152361 - 87196332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024550.1 9 - 10 1035 123
XM_017012871.1 XP_016868360.1 10 - 11 2051 123
XM_017012872.1 XP_016868361.1 9 - 10 1957 123
XM_017012873.1 XP_016868362.1 10 - 11 1846 123
XM_017012874.1 XP_016868363.1 10 - 11 1082 123
XM_017012877.1 XP_016868366.1 9 - 10 944 123
GenBank mRNA AF202144.1 - 9 - 10 984 123
AK126664.1 - 9 - 10 1530 123


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB306792.1 - 2 - 3 Chr. 7 - 87174622 - 87184414 [-] on Build GRCh38 123
GenBankmRNA AI609561.1 - 1 - 2 Chr. 7 - 87173601 - 87182162 [-] on Build GRCh38 124
GenBankmRNA BQ716241.1 - 302 Chr. - - [-] on Build 123


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101927420 LOC101927420 -24.6kb Anti-sense Chr. - 87151423 - 87152493 on Build GRCh38
Downstream transmembrane protein 243 TMEM243 C7orf23, MM-TRAG, MMTRAG 13.8kb Anti-sense Chr. - 87195593 - 87220603 on Build GRCh38

View Full Product Details