Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DMTF1
- Entrez Gene ID
-
9988
- Gene Name
- cyclin D binding myb like transcription factor 1
- Gene Aliases
-
DMP1, DMTF, MRUL, hDMP1
- Chromosome Location
-
Chr.7: 87152361 - 87196332 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB137203.1 |
- |
2 - 3 |
187 |
|
Chr. 7 - 87174641 - 87182340 [+] on Build GRCh38 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101927420 |
LOC101927420 |
|
-22.9kb
|
Anti-sense |
Chr. - 87151423 - 87152493 on Build GRCh38 |
Downstream |
transmembrane protein 243 |
TMEM243 |
C7orf23, MM-TRAG, MMTRAG |
14.8kb
|
Anti-sense |
Chr. - 87195593 - 87220603 on Build GRCh38 |
View Full Product Details