The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205316.1 | NP_001192245.1 | 2 - 3 | 582 | 63 |
GenBank mRNA | BC020600.1 | - | 2 - 3 | 557 | 63 |