The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161521.1 | NP_001154993.1 | 3 - 4 | 392 | 65 |
NM_007246.3 | NP_009177.3 | 3 - 4 | 515 | 65 | |
XM_011531572.2 | XP_011529874.1 | 4 - 5 | 505 | 65 | |
XM_017007674.1 | XP_016863163.1 | 4 - 5 | 465 | 65 | |
XM_017007675.1 | XP_016863164.1 | 2 - 3 | 230 | 65 | |
GenBank mRNA | AF059569.1 | - | 3 - 4 | 364 | 65 |
AK294103.1 | - | 3 - 4 | 386 | 65 | |
AK315372.1 | - | 3 - 4 | 360 | 65 | |
AL541403.3 | - | 3 - 4 | 515 | 65 | |
BC022503.1 | - | 3 - 4 | 497 | 65 | |
BC036468.2 | - | 3 - 4 | 456 | 65 | |
DA250099.1 | - | 3 - 4 | 392 | 65 |