The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005021.4 | NP_005012.2 | 9 - 10 | 1023 | 94 |
NR_133007.1 | 9 - 10 | 1023 | 94 | ||
XM_011535897.1 | XP_011534199.1 | 7 - 8 | 848 | 94 | |
XM_017010932.1 | XP_016866421.1 | 7 - 8 | 1068 | 94 | |
GenBank mRNA | AK024899.1 | - | 9 - 10 | 956 | 94 |
AK226107.1 | - | 10 - 11 | 1201 | 94 | |
AK292903.1 | - | 9 - 10 | 954 | 94 | |
AK309080.1 | - | 11 - 12 | 1475 | 94 | |
AK310691.1 | - | 8 - 9 | 1009 | 94 | |
AK315893.1 | - | 10 - 11 | 1211 | 94 | |
DQ234347.1 | - | 9 - 10 | 873 | 94 | |
EF560735.1 | - | 9 - 10 | 933 | 94 |