Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDNF
Entrez Gene ID
441549
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
ARMETL1
Chromosome Location
Chr.10: 14819245 - 14838073 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC144619.1 - 3 - 4 397 71

View Full Product Details