Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRAS1
Entrez Gene ID
80144
Gene Name
Fraser extracellular matrix complex subunit 1
Gene Aliases
-
Chromosome Location
Chr.4: 78057323 - 78544269 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166133.1 NP_001159605.1 5 - 6 905 72
NM_025074.6 NP_079350.5 5 - 6 905 72
XM_006714316.2 XP_006714379.1 5 - 6 1152 72
GenBank mRNA BC131820.1 - 5 - 6 495 72
BC131821.1 - 5 - 6 495 72

View Full Product Details