Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K15
Entrez Gene ID
389840
Gene Name
mitogen-activated protein kinase kinase kinase 15
Gene Aliases
ASK3, bA723P2.3
Chromosome Location
Chr.X: 19360058 - 19515459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001671.3 NP_001001671.3 10 - 11 1591 101
XM_011545507.2 XP_011543809.2 11 - 12 1818 101
XM_011545508.2 XP_011543810.2 11 - 12 1818 101
XM_011545510.2 XP_011543812.1 4 - 5 2331 101
XM_017029511.1 XP_016885000.1 11 - 12 1820 101
GenBank mRNA CN293540.1 - 4 - 5 603 101

View Full Product Details