The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014978.2 | NP_055793.1 | 19 - 20 | 2968 | 62 |
XM_011539542.2 | XP_011537844.1 | 14 - 15 | 1719 | 62 | |
GenBank mRNA | AB028982.1 | - | 19 - 20 | 2968 | 62 |
BC117417.1 | - | 19 - 20 | 2784 | 62 |