The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324312.1 | NP_001311241.1 | 3 - 4 | 506 | 78 |
NM_001324313.1 | NP_001311242.1 | 3 - 4 | 506 | 78 | |
NM_001324314.1 | NP_001311243.1 | 4 - 5 | 688 | 78 | |
NM_001324315.1 | NP_001311244.1 | 3 - 4 | 1363 | 78 | |
NM_152707.3 | NP_689920.1 | 3 - 4 | 506 | 78 | |
NR_136737.1 | 3 - 4 | 1363 | 78 | ||
XM_011540211.2 | XP_011538513.1 | 3 - 4 | 506 | 78 | |
XM_017016695.1 | XP_016872184.1 | 3 - 4 | 506 | 78 | |
GenBank mRNA | AK123772.1 | - | 3 - 4 | 1364 | 78 |
AK290255.1 | - | 3 - 4 | 435 | 78 | |
AK295287.1 | - | 3 - 4 | 471 | 78 | |
AK298510.1 | - | 4 - 5 | 581 | 78 | |
BC030266.1 | - | 3 - 4 | 457 | 78 | |
DA023888.1 | - | 3 - 4 | 506 | 78 | |
M31659.1 | - | 4 - 5 | 525 | 78 |