The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002558.3 | NP_002549.1 | 4 - 5 | 869 | 62 |
XM_006721529.2 | XP_006721592.1 | 4 - 5 | 709 | 62 | |
XM_011523896.2 | XP_011522198.1 | 5 - 6 | 1261 | 62 | |
XM_011523897.2 | XP_011522199.1 | 4 - 5 | 709 | 62 | |
XM_011523898.2 | XP_011522200.1 | 4 - 5 | 1373 | 62 | |
XM_011523899.2 | XP_011522201.1 | 4 - 5 | 1373 | 62 | |
XM_011523900.2 | XP_011522202.1 | 4 - 5 | 1373 | 62 | |
GenBank mRNA | AF020498.1 | - | 4 - 5 | 704 | 62 |
AK313264.1 | - | 4 - 5 | 668 | 62 | |
BC027949.1 | - | 4 - 5 | 655 | 62 | |
BC044657.1 | - | 4 - 5 | 644 | 62 | |
U45448.1 | - | 4 - 5 | 624 | 62 | |
X83688.1 | - | 4 - 5 | 601 | 62 |