Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNN2
Entrez Gene ID
3781
Gene Name
potassium calcium-activated channel subfamily N member 2
Gene Aliases
KCa2.2, SK2, SKCA2, SKCa 2, hSK2
Chromosome Location
Chr.5: 114056006 - 114496500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021614.3 NP_067627.2 2 - 3 1038 72
XM_011543387.1 XP_011541689.1 7 - 8 2321 72
XM_011543389.1 XP_011541691.1 6 - 7 2252 72
GenBank mRNA AF239613.1 - 2 - 3 1038 72
AF397175.1 - 2 - 3 611 72
AK289948.1 - 2 - 3 970 72
AY258141.1 - 2 - 3 584 72
BC117454.1 - 2 - 3 749 72
BC117456.1 - 2 - 3 749 72

View Full Product Details