Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNN2
Entrez Gene ID
3781
Gene Name
potassium calcium-activated channel subfamily N member 2
Gene Aliases
KCa2.2, SK2, SKCA2, SKCa 2, hSK2
Chromosome Location
Chr.5: 114056006 - 114496500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021614.3 NP_067627.2 3 - 4 1456 80
XM_011543387.1 XP_011541689.1 8 - 9 2739 80
XM_011543389.1 XP_011541691.1 7 - 8 2670 80
GenBank mRNA AF239613.1 - 3 - 4 1456 80
AK289948.1 - 3 - 4 1388 80
AY258141.1 - 3 - 4 1002 80
BC117454.1 - 3 - 4 1167 80
BC117456.1 - 3 - 4 1167 80

View Full Product Details