The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198150.2 | NP_937793.1 | 2 - 3 | 414 | 143 |
XM_005271904.3 | XP_005271961.1 | 2 - 3 | 427 | 143 | |
GenBank mRNA | AK303855.1 | - | 2 - 3 | 466 | 143 |
AK316253.1 | - | 2 - 3 | 434 | 143 | |
AL832711.1 | - | 1 - 2 | 97 | 143 | |
AY358596.1 | - | 2 - 3 | 374 | 143 | |
AY875939.1 | - | 2 - 3 | 256 | 143 | |
BC036047.1 | - | 2 - 3 | 276 | 143 | |
BC050392.1 | - | 2 - 3 | 373 | 143 | |
BC130329.1 | - | 2 - 3 | 352 | 143 | |
BC144161.1 | - | 2 - 3 | 352 | 143 |