Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSK
Entrez Gene ID
153642
Gene Name
arylsulfatase family member K
Gene Aliases
TSULF
Chromosome Location
Chr.5: 95555108 - 95605102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198150.2 NP_937793.1 7 - 8 1481 152
XM_005271904.3 XP_005271961.1 8 - 9 1545 152
GenBank mRNA AK303855.1 - 7 - 8 1533 152
AY358596.1 - 7 - 8 1441 152
AY875939.1 - 7 - 8 1323 152
BC050392.1 - 8 - 9 1491 152
BC130329.1 - 7 - 8 1419 152
BC144161.1 - 8 - 9 1478 152

View Full Product Details