The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300741.1 | NP_001287670.1 | 5 - 6 | 1130 | 96 |
NM_031438.3 | NP_113626.1 | 5 - 6 | 1184 | 96 | |
XM_005272095.1 | XP_005272152.1 | 5 - 6 | 1169 | 96 | |
XM_005272097.3 | XP_005272154.1 | 5 - 6 | 1130 | 96 | |
GenBank mRNA | AK098066.1 | - | 5 - 6 | 1153 | 96 |
AK304010.1 | - | 5 - 6 | 1118 | 96 | |
AL136592.1 | - | 5 - 6 | 1139 | 96 | |
BC026748.1 | - | 2 - 3 | 206 | 96 | |
BC041099.1 | - | 5 - 6 | 1168 | 96 |