Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FAM13A
- Entrez Gene ID
-
10144
- Gene Name
- family with sequence similarity 13 member A
- Gene Aliases
-
ARHGAP48, FAM13A1
- Chromosome Location
-
Chr.4: 88725954 - 89111398 on Build GRCh38
View Full Product Details