Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
Chromosome Location
Chr.4: 88725954 - 89111398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015045.2 NP_001015045.1 4 - 5 648 76
NM_001265578.1 NP_001252507.1 3 - 4 606 76
NM_001265579.1 NP_001252508.1 4 - 5 648 76
NM_001265580.1 NP_001252509.1 2 - 3 564 76
NM_014883.3 NP_055698.2 10 - 11 1500 76
XM_005262681.2 XP_005262738.1 9 - 10 1458 76
XM_005262682.2 XP_005262739.1 10 - 11 1599 76
XM_005262683.2 XP_005262740.1 10 - 11 1500 76
XM_005262684.3 XP_005262741.1 9 - 10 1279 76
XM_006714057.3 XP_006714120.1 7 - 8 775 76
XM_011531516.1 XP_011529818.1 11 - 12 1521 76
XM_011531517.1 XP_011529819.1 9 - 10 1416 76
XM_011531518.1 XP_011529820.1 11 - 12 1650 76
XM_011531519.2 XP_011529821.1 9 - 10 1209 76
XM_017007624.1 XP_016863113.1 9 - 10 1458 76
XM_017007625.1 XP_016863114.1 9 - 10 1119 76
XM_017007626.1 XP_016863115.1 7 - 8 753 76
XM_017007627.1 XP_016863116.1 9 - 10 1344 76
XM_017007628.1 XP_016863117.1 8 - 9 1237 76
XM_017007629.1 XP_016863118.1 10 - 11 3231 76
XM_017007630.1 XP_016863119.1 8 - 9 1144 76
XM_017007631.1 XP_016863120.1 9 - 10 3189 76
XM_017007632.1 XP_016863121.1 7 - 8 1083 76
XM_017007633.1 XP_016863122.1 9 - 10 3147 76
XM_017007634.1 XP_016863123.1 3 - 4 557 76
GenBank mRNA AB020721.2 - 4 - 5 365 76
AK027138.1 - 4 - 5 488 76
AK091787.1 - 3 - 4 503 76
AK091881.1 - 11 - 12 1727 76
AK296932.1 - 2 - 3 433 76
BC086875.1 - 3 - 4 447 76

View Full Product Details