Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
Chromosome Location
Chr.4: 88725954 - 89111398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015045.2 NP_001015045.1 9 - 10 1321 67
NM_001265578.1 NP_001252507.1 8 - 9 1279 67
NM_001265579.1 NP_001252508.1 9 - 10 1321 67
NM_001265580.1 NP_001252509.1 7 - 8 1237 67
NM_014883.3 NP_055698.2 15 - 16 2173 67
XM_005262681.2 XP_005262738.1 14 - 15 2131 67
XM_005262682.2 XP_005262739.1 15 - 16 2272 67
XM_005262683.2 XP_005262740.1 15 - 16 2173 67
XM_005262684.3 XP_005262741.1 14 - 15 1952 67
XM_006714057.3 XP_006714120.1 12 - 13 1448 67
XM_011531516.1 XP_011529818.1 16 - 17 2194 67
XM_011531517.1 XP_011529819.1 14 - 15 2089 67
XM_011531518.1 XP_011529820.1 16 - 17 2323 67
XM_011531519.2 XP_011529821.1 14 - 15 1882 67
XM_017007624.1 XP_016863113.1 14 - 15 2131 67
XM_017007625.1 XP_016863114.1 14 - 15 1792 67
XM_017007626.1 XP_016863115.1 12 - 13 1426 67
XM_017007627.1 XP_016863116.1 14 - 15 2017 67
XM_017007628.1 XP_016863117.1 13 - 14 1910 67
XM_017007629.1 XP_016863118.1 15 - 16 3904 67
XM_017007630.1 XP_016863119.1 13 - 14 1817 67
XM_017007631.1 XP_016863120.1 14 - 15 3862 67
XM_017007632.1 XP_016863121.1 12 - 13 1756 67
XM_017007633.1 XP_016863122.1 14 - 15 3820 67
XM_017007634.1 XP_016863123.1 8 - 9 1230 67
GenBank mRNA AB020721.2 - 9 - 10 1038 67
AK027138.1 - 9 - 10 1161 67
AK091328.1 - 2 - 3 932 67
AK091787.1 - 8 - 9 1439 67
AK296932.1 - 7 - 8 1106 67
BC086875.1 - 8 - 9 1120 67

View Full Product Details