The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022060.2 | NP_071343.2 | 6 - 7 | 1005 | 68 |
XM_005267986.4 | XP_005268043.1 | 6 - 7 | 1058 | 68 | |
GenBank mRNA | AK022878.1 | - | 6 - 7 | 1007 | 68 |
AK024790.1 | - | 2 - 3 | 297 | 68 | |
AK293198.1 | - | 5 - 6 | 784 | 68 | |
BC024779.1 | - | 6 - 7 | 953 | 68 |