The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261435.1 | NP_001248364.1 | 6 - 7 | 685 | 61 |
NM_001261436.1 | NP_001248365.1 | 6 - 7 | 685 | 61 | |
NM_139158.2 | NP_631897.1 | 6 - 7 | 595 | 61 | |
XM_005246781.3 | XP_005246838.1 | 6 - 7 | 685 | 61 | |
XM_005246782.4 | XP_005246839.1 | 7 - 8 | 932 | 61 | |
XM_011511650.2 | XP_011509952.1 | 6 - 7 | 685 | 61 | |
XM_011511652.1 | XP_011509954.1 | 6 - 7 | 685 | 61 | |
XM_011511654.2 | XP_011509956.1 | 6 - 7 | 685 | 61 | |
XM_011511655.2 | XP_011509957.1 | 3 - 4 | 692 | 61 | |
XM_011511656.2 | XP_011509958.1 | 3 - 4 | 299 | 61 | |
GenBank mRNA | AB053308.1 | - | 6 - 7 | 549 | 61 |
AK292434.1 | - | 7 - 8 | 802 | 61 | |
AK310050.1 | - | 8 - 9 | 918 | 61 | |
AX772791.1 | - | 6 - 7 | 549 | 61 | |
BC038807.1 | - | 6 - 7 | 643 | 61 | |
BX647394.1 | - | 6 - 7 | 649 | 61 |