The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261435.1 | NP_001248364.1 | 7 - 8 | 816 | 64 |
NM_001261436.1 | NP_001248365.1 | 7 - 8 | 816 | 64 | |
NM_139158.2 | NP_631897.1 | 7 - 8 | 726 | 64 | |
XM_005246781.3 | XP_005246838.1 | 7 - 8 | 816 | 64 | |
XM_005246782.4 | XP_005246839.1 | 8 - 9 | 1063 | 64 | |
XM_011511650.2 | XP_011509952.1 | 7 - 8 | 816 | 64 | |
XM_011511652.1 | XP_011509954.1 | 7 - 8 | 816 | 64 | |
XM_011511654.2 | XP_011509956.1 | 7 - 8 | 816 | 64 | |
XM_011511655.2 | XP_011509957.1 | 4 - 5 | 823 | 64 | |
XM_011511656.2 | XP_011509958.1 | 4 - 5 | 430 | 64 | |
XM_011511657.2 | XP_011509959.1 | 2 - 3 | 327 | 64 | |
GenBank mRNA | AB053308.1 | - | 7 - 8 | 680 | 64 |
AK131512.1 | - | 6 - 7 | 753 | 64 | |
AK292434.1 | - | 8 - 9 | 933 | 64 | |
AK310050.1 | - | 9 - 10 | 1049 | 64 | |
AX772791.1 | - | 7 - 8 | 680 | 64 | |
BC038807.1 | - | 7 - 8 | 774 | 64 | |
BX647394.1 | - | 7 - 8 | 780 | 64 |