Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APMAP
Entrez Gene ID
57136
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
BSCv, C20orf3
Chromosome Location
Chr.20: 24962925 - 24992974 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020531.2 NP_065392.1 4 - 5 526 60
XM_005260763.3 XP_005260820.1 4 - 5 667 60
GenBank mRNA AB033767.1 - 4 - 5 465 60
AK291129.1 - 4 - 5 487 60
AK301959.1 - 4 - 5 667 60
AY359076.1 - 3 - 4 313 60
BC003501.1 - 4 - 5 450 60
CF131759.1 - 4 - 5 495 60

View Full Product Details