The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019004.1 | NP_061877.1 | 3 - 4 | 857 | 65 |
XM_005250458.3 | XP_005250515.1 | 3 - 4 | 875 | 65 | |
XM_011516335.2 | XP_011514637.1 | 3 - 4 | 439 | 65 | |
XM_017012359.1 | XP_016867848.1 | 3 - 4 | 875 | 65 | |
GenBank mRNA | AB037807.1 | - | 3 - 4 | 857 | 65 |
BC063861.1 | - | 3 - 4 | 833 | 65 |