Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 92246216 - 92401773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019004.1 NP_061877.1 4 - 5 1044 80
XM_005250458.3 XP_005250515.1 4 - 5 1062 80
XM_011516335.2 XP_011514637.1 4 - 5 626 80
XM_017012359.1 XP_016867848.1 4 - 5 1062 80
GenBank mRNA AB037807.1 - 4 - 5 1044 80
AK025475.1 - 1 - 2 141 80
BC063861.1 - 4 - 5 1020 80

View Full Product Details