The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134999.1 | NP_001128471.1 | 1 - 2 | 175 | 98 |
NM_001135000.1 | NP_001128472.1 | 1 - 2 | 175 | 98 | |
NM_006832.2 | NP_006823.1 | 1 - 2 | 175 | 98 | |
XM_005267285.2 | XP_005267342.1 | 1 - 2 | 270 | 98 | |
XM_006720008.2 | XP_006720071.1 | 1 - 2 | 270 | 98 | |
XM_006720009.2 | XP_006720072.1 | 1 - 2 | 270 | 98 | |
XM_006720010.2 | XP_006720073.1 | 1 - 2 | 270 | 98 | |
GenBank mRNA | AF443279.1 | - | 1 - 2 | 70 | 98 |
AK291738.1 | - | 1 - 2 | 128 | 98 | |
BC017327.2 | - | 1 - 2 | 175 | 98 | |
BX161467.1 | - | 1 - 2 | 96 | 98 |