The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144999.2 | NP_001138471.1 | 27 - 28 | 2813 | 139 |
NM_001145000.2 | NP_001138472.1 | 25 - 26 | 2989 | 139 | |
NM_002210.4 | NP_002201.1 | 27 - 28 | 3097 | 139 | |
GenBank mRNA | AB209894.1 | - | 25 - 26 | 2972 | 139 |
AK302990.1 | - | 27 - 28 | 2813 | 139 | |
BC047454.1 | - | 2 - 3 | 181 | 139 | |
BC126231.1 | - | 27 - 28 | 2950 | 139 | |
BC136442.1 | - | 27 - 28 | 2950 | 139 | |
BC144100.1 | - | 25 - 26 | 2842 | 139 | |
EF560727.1 | - | 27 - 28 | 2865 | 139 | |
M14648.1 | - | 27 - 28 | 2862 | 139 |