The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144999.2 | NP_001138471.1 | 2 - 3 | 310 | 91 |
NM_001145000.2 | NP_001138472.1 | 2 - 3 | 594 | 91 | |
NM_002210.4 | NP_002201.1 | 2 - 3 | 594 | 91 | |
GenBank mRNA | AB209894.1 | - | 2 - 3 | 577 | 91 |
AK302550.1 | - | 2 - 3 | 594 | 91 | |
AK302990.1 | - | 2 - 3 | 310 | 91 | |
AK309711.1 | - | 2 - 3 | 594 | 91 | |
BC126231.1 | - | 2 - 3 | 447 | 91 | |
BC136442.1 | - | 2 - 3 | 447 | 91 | |
BC144100.1 | - | 2 - 3 | 447 | 91 | |
EF560727.1 | - | 2 - 3 | 362 | 91 | |
M14648.1 | - | 2 - 3 | 359 | 91 |