Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CECR2
Entrez Gene ID
27443
Gene Name
CECR2, histone acetyl-lysine reader
Gene Aliases
-
Chromosome Location
Chr.22: 17359949 - 17558155 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290046.1 NP_001276975.1 10 - 11 1190 111
NM_001290047.1 NP_001276976.1 10 - 11 1659 111
XM_011546128.2 XP_011544430.1 9 - 10 1263 111
XM_011546129.2 XP_011544431.1 10 - 11 1347 111
XM_011546132.2 XP_011544434.1 10 - 11 1347 111
XM_017028785.1 XP_016884274.1 10 - 11 1347 111
GenBank mRNA AB051527.1 - 2 - 3 208 111
AF336133.1 - 10 - 11 1719 111
AF411609.1 - 6 - 7 716 111
AK308330.1 - 10 - 11 1191 111

View Full Product Details