Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HDAC4
- Entrez Gene ID
-
9759
- Gene Name
- histone deacetylase 4
- Gene Aliases
-
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
- Chromosome Location
-
Chr.2: 239048168 - 239401647 on Build GRCh38
View Full Product Details