Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDAC4
Entrez Gene ID
9759
Gene Name
histone deacetylase 4
Gene Aliases
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
Chromosome Location
Chr.2: 239048168 - 239401647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006037.3 NP_006028.2 21 - 22 3430 91
XM_006712877.3 XP_006712940.1 20 - 21 9376 91
XM_006712878.3 XP_006712941.1 21 - 22 2950 91
XM_006712879.3 XP_006712942.1 20 - 21 8843 91
XM_006712880.3 XP_006712943.1 20 - 21 8855 91
XM_011512217.2 XP_011510519.1 20 - 21 9391 91
XM_011512218.2 XP_011510520.1 20 - 21 9376 91
XM_011512219.2 XP_011510521.1 20 - 21 9361 91
XM_011512220.2 XP_011510522.1 20 - 21 3130 91
XM_011512221.1 XP_011510523.1 21 - 22 3118 91
XM_011512222.2 XP_011510524.1 21 - 22 3419 91
XM_011512223.2 XP_011510525.1 21 - 22 2933 91
XM_011512224.2 XP_011510526.1 20 - 21 3321 91
XM_011512225.2 XP_011510527.1 20 - 21 3306 91
XM_011512226.2 XP_011510528.1 20 - 21 2878 91
XM_011512227.2 XP_011510529.1 19 - 20 8405 91
XM_011512230.1 XP_011510532.1 15 - 16 2316 91
XM_017005394.1 XP_016860883.1 19 - 20 9260 91
XM_017005395.1 XP_016860884.1 17 - 18 2182 91
GenBank mRNA AB006626.2 - 21 - 22 3430 91
AF132607.1 - 20 - 21 2638 91
AK302719.1 - 15 - 16 1999 91
BC039904.1 - 21 - 22 2697 91

View Full Product Details