The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014239.3 | NP_055054.1 | 3 - 4 | 514 | 66 |
GenBank mRNA | AF035280.1 | - | 3 - 4 | 478 | 66 |
BC000494.2 | - | 3 - 4 | 480 | 66 | |
BC003165.1 | - | 3 - 4 | 480 | 66 | |
BC011750.2 | - | 3 - 4 | 455 | 66 | |
BI908906.1 | - | 3 - 4 | 514 | 66 | |
BX161473.1 | - | 3 - 4 | 512 | 66 | |
L40395.1 | - | 3 - 4 | 416 | 66 |