Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
Chromosome Location
Chr.14: 75002909 - 75009591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014239.3 NP_055054.1 5 - 6 776 86
GenBank mRNA AF035280.1 - 5 - 6 740 86
BC000494.2 - 5 - 6 742 86
BC003165.1 - 5 - 6 742 86
BC011750.2 - 5 - 6 717 86
BX161473.1 - 5 - 6 774 86
L40395.1 - 5 - 6 678 87

View Full Product Details