Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LPIN1
- Entrez Gene ID
-
23175
- Gene Name
- lipin 1
- Gene Aliases
-
PAP1
- Chromosome Location
-
Chr.2: 11677544 - 11827409 on Build GRCh38
View Full Product Details